Tisk…

Anti-Aspartate beta hydroxylase Rabbit Monoclonal Antibody [clone: EPR10351]

Dodavatel: Abcam
AB172475-100 AB172475-1ML
ABCAAB172475-100EA 20130 CZK
ABCAAB172475-100 ABCAAB172475-1ML
Anti-Aspartate beta hydroxylase Rabbit Monoclonal Antibody [clone: EPR10351]
Protilátky
Rabbit monoclonal [EPR10351] to Aspartate beta hydroxylase.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Aspartate beta hydroxylase
Clonality: Monoclonal
Clone: EPR10351
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR