Tisk…
Anti-PTHLH Rabbit Monoclonal Antibody [clone: EPR26606-236]
Dodavatel: Abcam
AB314119-100UG AB314119-1MG
ABCAAB314119-100UGEA 29040 CZK
ABCAAB314119-100UG ABCAAB314119-1MG
Anti-PTHLH Rabbit Monoclonal Antibody [clone: EPR26606-236]
Protilátky
Rabbit monoclonal [EPR26606-236] to PTHLH - BSA and Azide free.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: PTHLH
Clonality: Monoclonal
Clone: EPR26606-236
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human