Tisk…

Anti-IgG Goat Polyclonal Antibody (Gold)

Dodavatel: Abcam
AB270557-1 AB270557-200
ABCAAB270557-1EA 13190 CZK
ABCAAB270557-1 ABCAAB270557-200
Anti-IgG Goat Polyclonal Antibody (Gold)
Protilátky
Goat Anti-Mouse IgG (20nm Gold, 10 OD).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR