Tisk…

Anti-CD7 Rabbit Monoclonal Antibody [clone: EPR4242]

Dodavatel: Abcam
AB230834-100 AB230834-1
ABCAAB230834-100EA 29040 CZK
ABCAAB230834-100 ABCAAB230834-1
Anti-CD7 Rabbit Monoclonal Antibody [clone: EPR4242]
Protilátky
Rabbit monoclonal [EPR4242] to CD7 - BSA and Azide free.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: EPR4242
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR