Tisk…

Anti-Glucose Transporter GLUT4 Rabbit Monoclonal Antibody [clone: EPR26571-45]

Dodavatel: Abcam
AB313776-100UG AB313776-1MG
ABCAAB313776-100UGEA 29040 CZK
ABCAAB313776-100UG ABCAAB313776-1MG
Anti-Glucose Transporter GLUT4 Rabbit Monoclonal Antibody [clone: EPR26571-45]
Protilátky
Rabbit monoclonal [EPR26571-45] to Glucose Transporter GLUT4 - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glucose Transporter GLUT4
Clonality: Monoclonal
Clone: EPR26571-45
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR