Tisk…

Anti-Bcl6 Rabbit Monoclonal Antibody [clone: SP155]

Dodavatel: Abcam
AB243920-100 AB243920-1
ABCAAB243920-100EA 29040 CZK
ABCAAB243920-100 ABCAAB243920-1
Anti-Bcl6 Rabbit Monoclonal Antibody [clone: SP155]
Protilátky
Rabbit monoclonal [SP155] to Bcl6 - BSA and Azide free.

Type: Primary
Antigen: Bcl6
Clonality: Monoclonal
Clone: SP155
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR