Tisk…
Anti-Glucose 6 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR24680-118]
Dodavatel: Abcam
AB303690-1 AB303690-100
ABCAAB303690-1EA 248130 CZK
ABCAAB303690-1 ABCAAB303690-100
Anti-Glucose 6 Phosphate Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR24680-118]
Protilátky
Rabbit monoclonal [EPR24680-118] to Glucose 6 Phosphate Dehydrogenase - BSA and Azide free (Detector).
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Glucose 6 Phosphate Dehydrogenase
Clonality: Monoclonal
Clone: EPR24680-118
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human