Tisk…

Anti-ENT1 Rabbit Monoclonal Antibody [clone: SP120]

Dodavatel: Abcam
AB182023-100 AB182023-500 AB182023-1
ABCAAB182023-100EA 20310 CZK
ABCAAB182023-100 ABCAAB182023-500 ABCAAB182023-1
Anti-ENT1 Rabbit Monoclonal Antibody [clone: SP120]
Protilátky
Rabbit monoclonal [SP120] to ENT1.

Type: Primary
Antigen: ENT1
Clonality: Monoclonal
Clone: SP120
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR