Tisk…

Anti-ACTH Mouse Monoclonal Antibody [clone: AH26]

Dodavatel: Abcam
AB212734-100 AB212734-1MG
ABCAAB212734-100EA 29040 CZK
ABCAAB212734-100 ABCAAB212734-1MG
Anti-ACTH Mouse Monoclonal Antibody [clone: AH26]
Protilátky
Mouse monoclonal [AH26] to ACTH - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ACTH
Clonality: Monoclonal
Clone: AH26
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR