Tisk…
Anti-NEK2 Mouse Monoclonal Antibody [clone: 20/Nek2]
Dodavatel: Abcam
AB279717-100 AB279717-1MG AB279717-20UG
ABCAAB279717-100EA 20310 CZK
ABCAAB279717-100 ABCAAB279717-1MG ABCAAB279717-20UG
Anti-NEK2 Mouse Monoclonal Antibody [clone: 20/Nek2]
Protilátky
Mouse monoclonal [20/NEK2] to NEK2.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: NEK2
Clonality: Monoclonal
Clone: 20/Nek2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human