Tisk…

Anti-RBPJK Rat Monoclonal Antibody [clone: 7A11]

Dodavatel: Abcam
AB252858-100 AB252858-1MG
ABCAAB252858-100EA 21740 CZK
ABCAAB252858-100 ABCAAB252858-1MG
Anti-RBPJK Rat Monoclonal Antibody [clone: 7A11]
Protilátky
Rat monoclonal [7A11] to RBPJK.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RBPJK
Clonality: Monoclonal
Clone: 7A11
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR