Tisk…

Anti-Lamin B2 Rabbit Monoclonal Antibody [clone: EPR9701(B)]

Dodavatel: Abcam
AB151735-100 AB151735-1ML
ABCAAB151735-100EA 20850 CZK
ABCAAB151735-100 ABCAAB151735-1ML
Anti-Lamin B2 Rabbit Monoclonal Antibody [clone: EPR9701(B)]
Protilátky
Rabbit monoclonal [EPR9701(B)] to Lamin B2.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Lamin B2
Clonality: Monoclonal
Clone: EPR9701(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR