Tisk…
Anti-ICA 512/PTPRN Rabbit Monoclonal Antibody [clone: EPR20718-111]
Dodavatel: Abcam
AB244950-100 AB244950-1
ABCAAB244950-100EA 26720 CZK
ABCAAB244950-100 ABCAAB244950-1
Anti-ICA 512/PTPRN Rabbit Monoclonal Antibody [clone: EPR20718-111]
Protilátky
Rabbit monoclonal [EPR20718-111] to ICA 512/PTPRN - BSA and Azide free (Detector).
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ICA 512/PTPRN
Clonality: Monoclonal
Clone: EPR20718-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human