Tisk…

Anti-SELO Rabbit Monoclonal Antibody [clone: EPR11968]

Dodavatel: Abcam
AB172957-100 AB172957-10 AB172957-1ML
ABCAAB172957-100EA 19240 CZK
ABCAAB172957-100 ABCAAB172957-10 ABCAAB172957-1ML
Anti-SELO Rabbit Monoclonal Antibody [clone: EPR11968]
Protilátky
Rabbit monoclonal [EPR11968] to SELO.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SELO
Clonality: Monoclonal
Clone: EPR11968
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR