Tisk…

Anti-VEGF Receptor 2 Rabbit Monoclonal Antibody [clone: EPR21884-236]

Dodavatel: Abcam
AB233693-100 AB233693-20UL AB233693-1ML
ABCAAB233693-100EA 19060 CZK
ABCAAB233693-100 ABCAAB233693-20UL ABCAAB233693-1ML
Anti-VEGF Receptor 2 Rabbit Monoclonal Antibody [clone: EPR21884-236]
Protilátky
Rabbit monoclonal [EPR21884-236] to VEGF Receptor 2.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: VEGF Receptor 2
Clonality: Monoclonal
Clone: EPR21884-236
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR