Tisk…

Anti-NEK7 Rabbit Monoclonal Antibody [clone: EPR4900]

Dodavatel: Abcam
AB240065-100 AB240065-1
ABCAAB240065-100EA 29040 CZK
ABCAAB240065-100 ABCAAB240065-1
Anti-NEK7 Rabbit Monoclonal Antibody [clone: EPR4900]
Protilátky
Rabbit monoclonal [EPR4900] to NEK7 - BSA and Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NEK7
Clonality: Monoclonal
Clone: EPR4900
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR