Tisk…
Anti-Bad Rabbit Monoclonal Antibody [clone: EPR18375-130]
Dodavatel: Abcam
AB244638-100 AB244638-1
ABCAAB244638-100EA 26720 CZK
ABCAAB244638-100 ABCAAB244638-1
Anti-Bad Rabbit Monoclonal Antibody [clone: EPR18375-130]
Protilátky
Rabbit monoclonal [EPR18375-130] to Bad - BSA and Azide free (Capture).
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Bad
Clonality: Monoclonal
Clone: EPR18375-130
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse