Tisk…
Anti-CRISPR-Cas9 Rabbit Monoclonal Antibody [clone: EPR18991]
Dodavatel: Abcam
AB189380-100 AB189380-1ML
ABCAAB189380-100EA 19420 CZK
ABCAAB189380-100 ABCAAB189380-1ML
Anti-CRISPR-Cas9 Rabbit Monoclonal Antibody [clone: EPR18991]
Protilátky
Rabbit monoclonal [EPR18991] to CRISPR-Cas9.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CRISPR-Cas9
Clonality: Monoclonal
Clone: EPR18991
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat