Tisk…

Anti-Troponin I fast skeletal muscle Rabbit Monoclonal Antibody [clone: EPR14927(B)]

Dodavatel: Abcam
AB184554-100 AB184554-1ML
ABCAAB184554-100EA 19060 CZK
ABCAAB184554-100 ABCAAB184554-1ML
Anti-Troponin I fast skeletal muscle Rabbit Monoclonal Antibody [clone: EPR14927(B)]
Protilátky
Rabbit monoclonal [EPR14927(B)] to Troponin I fast skeletal muscle.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Troponin I fast skeletal muscle
Clonality: Monoclonal
Clone: EPR14927(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR