Tisk…

Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5] (Alexa Fluor® 488)

Dodavatel: Abcam
AB309369-100UL AB309369-10UL
ABCAAB309369-100ULEA 23340 CZK
ABCAAB309369-100UL ABCAAB309369-10UL
Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5] (Alexa Fluor® 488)
Protilátky
Alexa Fluor® 488 Mouse monoclonal [TAL 1B5] to HLA-DR.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: TAL 1B5
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR