Tisk…

Anti-Acid sphingomyelinase Rabbit Monoclonal Antibody [clone: EPR23090-181]

Dodavatel: Abcam
AB272729-100 AB272729-1ML
ABCAAB272729-100EA 20850 CZK
ABCAAB272729-100 ABCAAB272729-1ML
Anti-Acid sphingomyelinase Rabbit Monoclonal Antibody [clone: EPR23090-181]
Protilátky
Rabbit monoclonal [EPR23090-181] to Acid sphingomyelinase.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Acid sphingomyelinase
Clonality: Monoclonal
Clone: EPR23090-181
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR