Tisk…

Anti-MeCP2 Mouse Monoclonal Antibody [clone: 4B4]

Dodavatel: Abcam
AB255834-100 AB255834-1
ABCAAB255834-100EA 29040 CZK
ABCAAB255834-100 ABCAAB255834-1
Anti-MeCP2 Mouse Monoclonal Antibody [clone: 4B4]
Protilátky
Mouse monoclonal [4B4] to MeCP2 - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MeCP2
Clonality: Monoclonal
Clone: 4B4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR