Tisk…

Anti-Collagen VI Rabbit Monoclonal Antibody [clone: EPR7888(N)]

Dodavatel: Abcam
AB172606-100 AB172606-1ML
ABCAAB172606-100EA 19420 CZK
ABCAAB172606-100 ABCAAB172606-1ML
Anti-Collagen VI Rabbit Monoclonal Antibody [clone: EPR7888(N)]
Protilátky
Rabbit monoclonal [EPR7888(N)] to Collagen VI.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Collagen VI
Clonality: Monoclonal
Clone: EPR7888(N)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR