Tisk…
Anti-COX2 / Cyclooxygenase 2 Mouse Monoclonal Antibody [clone: 33/Cox-2 H1L1]
Dodavatel: Abcam
AB300668-100 AB300668-10 AB300668-20UG AB300668-1MG
ABCAAB300668-100EA 13540 CZK
ABCAAB300668-100 ABCAAB300668-10 ABCAAB300668-20UG ABCAAB300668-1MG
Anti-COX2 / Cyclooxygenase 2 Mouse Monoclonal Antibody [clone: 33/Cox-2 H1L1]
Protilátky
Mouse monoclonal [33/Cox-2] to COX2 / Cyclooxygenase 2.
Type: Primary
Antigen: COX2 / Cyclooxygenase 2
Clonality: Monoclonal
Clone: 33/Cox-2 H1L1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat