Tisk…
Anti-IL-4 Rabbit Monoclonal Antibody [clone: EPR22879-236]
Dodavatel: Abcam
AB225638-100 AB225638-1ML
ABCAAB225638-100EA 0 CZK
ABCAAB225638-100 ABCAAB225638-1ML
Anti-IL-4 Rabbit Monoclonal Antibody [clone: EPR22879-236]
Protilátky
Rabbit monoclonal [EPR22879-236] to IL-4.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: EPR22879-236
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse