Tisk…

Anti-delta 1 Catenin/CAS (phospho Y280) Rabbit Monoclonal Antibody [clone: EPR2384(2)]

Dodavatel: Abcam
AB173578-100 AB173578-1ML
ABCAAB173578-100EA 18890 CZK
ABCAAB173578-100 ABCAAB173578-1ML
Anti-delta 1 Catenin/CAS (phospho Y280) Rabbit Monoclonal Antibody [clone: EPR2384(2)]
Protilátky
Rabbit monoclonal [EPR2384(2)] to delta 1 Catenin/CAS (phospho Y280).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: delta 1 Catenin/CAS
Clonality: Monoclonal
Clone: EPR2384(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR