Tisk…

Anti-ASAH1 Mouse Monoclonal Antibody [clone: 23Acid Ceramidase]

Dodavatel: Abcam
AB302633-100 AB302633-20UG AB302633-1MG
ABCAAB302633-100EA 20130 CZK
ABCAAB302633-100 ABCAAB302633-20UG ABCAAB302633-1MG
Anti-ASAH1 Mouse Monoclonal Antibody [clone: 23Acid Ceramidase]
Protilátky
Mouse monoclonal [23/Acid Ceramidase] to ASAH1.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ASAH1
Clonality: Monoclonal
Clone: 23Acid Ceramidase
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR