Tisk…

Anti-IL-33 Rabbit Monoclonal Antibody [clone: EPR20417]

Dodavatel: Abcam
AB236036-100 AB236036-1
ABCAAB236036-100EA 29040 CZK
ABCAAB236036-100 ABCAAB236036-1
Anti-IL-33 Rabbit Monoclonal Antibody [clone: EPR20417]
Protilátky
Rabbit monoclonal [EPR20417] to IL-33 - BSA and Azide free.

Type: Primary
Antigen: IL-33
Clonality: Monoclonal
Clone: EPR20417
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR