Tisk…

Anti-MUC1 Mouse Monoclonal Antibody [clone: HMFG1 (aka 1.10.F3)]

Dodavatel: Abcam
AB230298-100 AB230298-1
ABCAAB230298-100EA 0 CZK
ABCAAB230298-100 ABCAAB230298-1
Anti-MUC1 Mouse Monoclonal Antibody [clone: HMFG1 (aka 1.10.F3)]
Protilátky
Mouse monoclonal [HMFG1 (aka 1.10.F3)] to MUC1 - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: HMFG1 (aka 1.10.F3)
Conjugation:
Epitope: This antibody recognizes a peptide epitope (PDTR) within the VNTR region of the extracellular domain of MUC1 (PubMed ID: PMC3021526).
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR