Tisk…

Anti-Polymeric immunoglobulin receptor/PIGR Rabbit Monoclonal Antibody [clone: EPR23314-256]

Dodavatel: Abcam
AB275030-100 AB275030-1
ABCAAB275030-100EA 29040 CZK
ABCAAB275030-100 ABCAAB275030-1
Anti-Polymeric immunoglobulin receptor/PIGR Rabbit Monoclonal Antibody [clone: EPR23314-256]
Protilátky
Rabbit monoclonal [EPR23314-256] to Polymeric immunoglobulin receptor/PIGR - BSA and Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Polymeric immunoglobulin receptor/PIGR
Clonality: Monoclonal
Clone: EPR23314-256
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR