Tisk…

Anti-Oligodendrocyte Specific Protein Rabbit Monoclonal Antibody [clone: EPR12726]

Dodavatel: Abcam
AB175236-100 AB175236-1ML
ABCAAB175236-100EA 13540 CZK
ABCAAB175236-100 ABCAAB175236-1ML
Anti-Oligodendrocyte Specific Protein Rabbit Monoclonal Antibody [clone: EPR12726]
Protilátky
Rabbit monoclonal [EPR12726] to Oligodendrocyte Specific Protein - Oligodendrocyte Marker.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Oligodendrocyte Specific Protein
Clonality: Monoclonal
Clone: EPR12726
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR