Tisk…

Anti-P2X3 Rabbit Monoclonal Antibody [clone: EPR26568-3]

Dodavatel: Abcam
AB300494-1 AB300494-100
ABCAAB300494-1EA 260960 CZK
ABCAAB300494-1 ABCAAB300494-100
Anti-P2X3 Rabbit Monoclonal Antibody [clone: EPR26568-3]
Protilátky
Rabbit monoclonal [EPR26568-3] to P2X3 - BSA and Azide free.

Type: Primary
Antigen: P2X3
Clonality: Monoclonal
Clone: EPR26568-3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR