Tisk…

Anti-KMT3A/HYPB/HIF-1 Rabbit Monoclonal Antibody [clone: EPR20927-67]

Dodavatel: Abcam
AB239350-100 AB239350-1ML
ABCAAB239350-100EA 21740 CZK
ABCAAB239350-100 ABCAAB239350-1ML
Anti-KMT3A/HYPB/HIF-1 Rabbit Monoclonal Antibody [clone: EPR20927-67]
Protilátky
Rabbit monoclonal [EPR20927-67] to KMT3A/HYPB/HIF-1.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: KMT3A/HYPB/HIF-1
Clonality: Monoclonal
Clone: EPR20927-67
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR