Tisk…

Anti-Alkaline Phosphatase Rabbit Monoclonal Antibody [clone: EPR20428-111]

Dodavatel: Abcam
AB252640-1 AB252640-100
ABCAAB252640-1EA 265410 CZK
ABCAAB252640-1 ABCAAB252640-100
Anti-Alkaline Phosphatase Rabbit Monoclonal Antibody [clone: EPR20428-111]
Protilátky
Rabbit monoclonal [EPR20428-111] to Alkaline Phosphatase - BSA and Azide free (Detector).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Alkaline Phosphatase
Clonality: Monoclonal
Clone: EPR20428-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR