Tisk…

Anti-Tau Rabbit Monoclonal Antibody [clone: SP70]

Dodavatel: Abcam
AB236219-100 AB236219-1
ABCAAB236219-100EA 29040 CZK
ABCAAB236219-100 ABCAAB236219-1
Anti-Tau Rabbit Monoclonal Antibody [clone: SP70]
Protilátky
Rabbit monoclonal [SP70] to Tau - BSA and Azide free.

Type: Primary
Antigen: Tau
Clonality: Monoclonal
Clone: SP70
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR