Tisk…

Anti-Bcl-2 Rat Monoclonal Antibody [clone: NOR 235J]

Dodavatel: Abcam
AB241548-100 AB241548-1MG
ABCAAB241548-100EA 20490 CZK
ABCAAB241548-100 ABCAAB241548-1MG
Anti-Bcl-2 Rat Monoclonal Antibody [clone: NOR 235J]
Protilátky
Rat monoclonal [NOR 235J] to Bcl-2.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: NOR 235J
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR