Tisk…

Anti-Leukotriene A4 hydrolase/LTA4H Rabbit Monoclonal Antibody [clone: EPR5713]

Dodavatel: Abcam
AB240064-100 AB240064-1
ABCAAB240064-100EA 29040 CZK
ABCAAB240064-100 ABCAAB240064-1
Anti-Leukotriene A4 hydrolase/LTA4H Rabbit Monoclonal Antibody [clone: EPR5713]
Protilátky
Rabbit monoclonal [EPR5713] to Leukotriene A4 hydrolase/LTA4H - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Leukotriene A4 hydrolase/LTA4H
Clonality: Monoclonal
Clone: EPR5713
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR