Tisk…
Separating funnels, Gilson
Dodavatel: SciLabware
443054N 443038N 443042N 443047N 443060N 443065N 443043N 443055N 443061N 443066N 443074N 443048N
BIBB443054NEA 4420 CZK
BIBB443054N BIBB443038N BIBB443042N 201-0070 BIBB443060N BIBB443065N BIBB443043N BIBB443055N BIBB443061N BIBB443066N BIBB443074N BIBB443048N
Separating funnels, Gilson
Nálevky Nálevky dělící
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.