Ammonium 1-pyrrolidinecarbodithioate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
M-Clarity™ quality level = MQ200
P8765-100G
P8765-1G
P8765-25G
P8765-5G
SIALP8765-100GEA
18450
CZK
SIALP8765-100G
SIALP8765-1G
SIALP8765-25G
SIALP8765-5G
Ammonium 1-pyrrolidinecarbodithioate, Sigma-Aldrich®
Ammonium 1-pyrrolidinecarbodithioate
Vzorec:
C₅H₁₂N₂S₂ |
MDL Number:
MFCD00012720 CAS číslo: 5108-96-3 EINECS: 225-834-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...