Ammonium 1-pyrrolidinecarbodithioate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
MQ200
09935-100G
09935-1KG
09935-25G
SIAL09935-100GEA
5570
CZK
SIAL09935-100G
SIAL09935-1KG
SIAL09935-25G
Ammonium 1-pyrrolidinecarbodithioate, Sigma-Aldrich®
Ammonium 1-pyrrolidinecarbodithioate
Vzorec:
C₅H₁₂N₂S₂ MW: 146.3 · 18.0 g/mol |
MDL Number:
MFCD00012720 CAS číslo: 5108-96-3 EINECS: 225-834-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...