Ammonium 1-pyrrolidinecarbodithioate for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
M-Clarity™ quality level = MQ200
8.21100.0025
8.21100.0100
8.21100.0025EA
1380
CZK
8.21100.0025
8.21100.0100
Ammonium 1-pyrrolidinecarbodithioate for synthesis, Sigma-Aldrich®
Ammonium 1-pyrrolidinecarbodithioate
Vzorec:
C₅H₁₂N₂S₂ MW: 146.3 · 18.0 g/mol |
MDL Number:
MFCD00012720 CAS číslo: 5108-96-3 EINECS: 225-834-4 |
Výsledky specifikačního testu
Solubility | 190 g/l (20 °C) |
Melting Point | 146 - 151 °C |
Molar Mass | 164.28 g/mol |
Bulk Density | 400 kg/m3 |
pH | 6 - 7 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...