Methyl-2-oxocyclopentanecarboxylate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methyl 2-cyclopentanonecarboxylate
M-Clarity™ quality level = MQ100
168106-100G
168106-25G
SIAL168106-100GEA
8190
CZK
SIAL168106-100G
SIAL168106-25G
Methyl-2-oxocyclopentanecarboxylate, Sigma-Aldrich®
Methyl-2-oxocyclopentanecarboxylate
Vzorec:
C₇H₁₀O₃ MW: 142,16 g/mol |
MDL Number:
MFCD00001411 CAS číslo: 10472-24-9 EINECS: 233-962-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...