Tisk…

Methyl-4-(bromomethyl)benzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 4-(bromomethyl)benzoate

M-Clarity™ quality level = MQ100

348155-25G 348155-5G
SIAL348155-25GEA 3860 CZK
SIAL348155-25G SIAL348155-5G
Methyl-4-(bromomethyl)benzoate, Sigma-Aldrich®
Methyl-4-(bromomethyl)benzoate
Vzorec: C₉H₉BrO₂
MW: 229,07 g/mol
MDL Number: MFCD00032453
CAS číslo: 2417-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR