Tisk…

Methyl-4-(bromomethyl)benzoate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Methyl 4-(bromomethyl)benzoate

A10557.06 A10557.14 A10557.22
A10557.06EA 2120 CZK
A10557.06 A10557.14 A10557.22
Methyl-4-(bromomethyl)benzoate ≥98%
Methyl-4-(bromomethyl)benzoate

Purity: 98% CAS No.: 2417-72-3 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Melting Point: 53-58° Boiling Point: 130-135°/2mm UN No.: 1759 MDL No.: MFCD00032453

Vzorec: C₉H₉BrO₂
MW: 229,07 g/mol
Bod varu: 130…135 °C (2 mmHg)
Bod tání: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00032453
CAS číslo: 2417-72-3
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR