2-(Benzyloxy)benzaldehyde ≥98%
Dodavatel: Thermo Fisher Scientific
L12866.09
L12866.18
L12866.09EA
1610
CZK
L12866.09
L12866.18
2-(Benzyloxy)benzaldehyde ≥98%
2-(Benzyloxy)benzaldehyde
Purity: 98%
CAS No.: 5896-17-3
Molecular Formula: C14H12O2
Formula Weight: 212.25
Melting Point: 46-47°
Beilstein Reference: 780307
Vzorec:
C₁₄H₁₂O₂ Bod tání: 46…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016583 CAS číslo: 5896-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...