tert-Butyldimethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
t-Butyldimethylsilyl trifluoromethanesulphonate
M-Clarity™ quality level = MQ200
226149-1G
226149-25G
226149-5G
SIAL226149-1GEA
1170
CZK
SIAL226149-1G
SIAL226149-25G
SIAL226149-5G
tert-Butyldimethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
tert-Butyldimethylsilyl trifluoromethanesulphonate
Vzorec:
C₇H₁₅F₃O₃SSi MW: 264,34 g/mol |
MDL Number:
MFCD00000405 CAS číslo: 69739-34-0 EINECS: 274-102-0 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...