EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate
A13724.09
A13724.18
A13724.30
A13724.09EA
1590
CZK
A13724.09
A13724.18
A13724.30
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) ≥99%
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
Purity: 99%
CAS No.: 16357-59-8
Molecular Formula: C14H17NO3
Formula Weight: 247.29
Storage Temperature: 2°C to 8°C
Melting Point: 63-66°
Boiling Point: 125-128°/0.1mm
Beilstein Reference: 533048
Merck Reference: 14,3518
MDL No.: MFCD00006703
Vzorec:
C₁₄H₁₇NO₃ MW: 247,29 g/mol Bod varu: 125…128 °C (0,1 mmHg) Bod tání: 63…66 °C |
MDL Number:
MFCD00006703 CAS číslo: 16357-59-8 EINECS: 240-418-2 Merck Index: 14,3518 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...