Tisk…

1,2-Benzisoxazole 95%

Dodavatel: Thermo Scientific
Varování

B23400.03 B23400.06
B23400.03EA 1120 CZK
B23400.03 B23400.06
1,2-Benzisoxazole 95%
1,2-Benzisoxazole

1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).


Purity: 97% CAS No.: 271-95-4 Molecular Formula: C7H5NO Formula Weight: 119.12 Storage Temperature: 2°C to 8°C Flash Point: 86°(186°F) Refractive Index: 1.5620 Boiling Point: 90-92°/15mm Beilstein Reference: 2154 MDL No.: MFCD00005852

Vzorec: C₇H₅NO
MW: 119,12 g/mol
Bod varu: 90…92 °C (15 mmHg)
Hustota: 1,169
Bod vzplanutí: 86 °C (186 °F)
MDL Number: MFCD00005852
CAS číslo: 271-95-4
EINECS: 205-983-1

Objednat

Výsledky specifikačního testu

Appearance (Color) Clear colorless to pale yellow
Form Liquid
Assay (GC) ≥94.0%
Refractive Index 1.5590-1.5650 @ 20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR