Tisk…
D-(-)-Isoascorbic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Erythorbic acid, (R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, D-Isoascorbic acid
M-Clarity™ quality level = MQ200
856061-100G 856061-500G
SIAL856061-100GEA 1290 CZK
SIAL856061-100G SIAL856061-500G
D-(-)-Isoascorbic acid, Sigma-Aldrich®
D-(-)-Isoascorbic acid
Vzorec: C₆H₈O₆
MW: 176,12 g/mol
MDL Number: MFCD00005378
CAS číslo: 89-65-6
EINECS: 201-928-0