Tisk…

D-(-)-Isoascorbic acid ≥98% (by titrimetric analysis)

Dodavatel: Thermo Fisher Scientific

36366.22 36366.36
36366.22EA 1700 CZK
36366.22 36366.36
D-(-)-Isoascorbic acid ≥98% (by titrimetric analysis)
D-(-)-Isoascorbic acid

Purity: 99% CAS No.: 89-65-6 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Odour: Odourless Merck Reference: 14,5126 MDL No.: MFCD00005378

Vzorec: C₆H₈O₆
Storage Temperature: Ambient
MDL Number: MFCD00005378
CAS číslo: 89-65-6
EINECS: 201-928-0
Merck Index: 14,5126

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR